Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.52415

Sigma-Aldrich

N3-Lys(Fmoc)-OH

Novabiochem®

Synonym(s):

N3-Lys(Fmoc)-OH, (s)-α-azido-ε-(Fmoc-amino)-hexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

azide

storage temp.

15-25°C

General description

A useful tool for the synthesis of branched, side-chain modified, cyclic peptides and peptide tools for click chemistry by Fmoc SPPS. The side-chain azido group is completely stable to piperidine and TFA, but can be readily converted to an amine on the solid phase or in solution by reduction with thiols or phosphines.

Associated Protocols and Technical Articles
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Literature references

[1] M. Meldal, et al. (1997) Tetrahedron Lett., 38, 2531.
[2] J. T. Lundquist & J. C. Pelletier (2001) Org. Lett., 3, 781.
[3] N. Nepomniaschiy, et al. (2008) Org. Lett., 10, 5243.
Novabiochem®

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (HPLC): ≥ 98.0 % (a/a)
Purity (TLC): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clear soluble
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service