Skip to Content
MilliporeSigma

8.52337

Fmoc-Phe(bis-Boc-4-guanidino)-OH

Novabiochem®

Synonym(s):

Fmoc-Phe(bis-Boc-4-guanidino)-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H40N4O8
CAS Number:
Molecular Weight:
644.71
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥90.0% (acidimetric), ≥93.0% (HPLC), ≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Boc, Fmoc

storage temp.

15-25°C

SMILES string

N([C@@H](Cc4ccc(cc4)N\C(=N/C(=O)OC(C)(C)C)\NC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C35H40N4O8/c1-34(2,3)46-32(43)38-30(39-33(44)47-35(4,5)6)36-22-17-15-21(16-18-22)19-28(29(40)41)37-31(42)45-20-27-25-13-9-7-11-23(25)24-12-8-10-14-26(24)27/h7-18,27-28H,19-20H2,1-6H3,(H,37,42)(H,40,41)(H2,36,38,39,43,44)/t28-/m0/s1

InChI key

CIEGZABLYKOZLG-NDEPHWFRSA-N

General description

A novel arginine mimetic.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -8.0 - -4.0 °
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 93.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 1.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library