Skip to Content
MilliporeSigma

8.52267

Fmoc-hArg(Pbf)-OH

Novabiochem®

Synonym(s):

Fmoc-hArg(Pbf)-OH, N-α-Fmoc-N G-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-homoarginine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H42N4O7S
CAS Number:
Molecular Weight:
662.80
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥95.0% (HPLC), ≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(\N=C(\NCCCC[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O)/N)c1c(c2c(c(c1C)C)OC(C2)(C)C)C

InChI key

DOGZBRBJANHMLA-LJAQVGFWSA-N

General description

Fmoc-homoArg(Pbf)-OH is a useful tool for the introduction of homoarginine during Fmoc SPPS. It can be used in exactly the same manner as Fmoc-Arg(Pbf)-OH.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -7.0 - -3.0 °
Purity (TLC(157B)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library