Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52110

Sigma-Aldrich

Fmoc-(Dmb)Gly-OH

Novabiochem®

Synonym(s):

Fmoc-(Dmb)Gly-OH, N-α-Fmoc-N-α-(2, 4-dimethoxybenzyl)-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25NO6
CAS Number:
Molecular Weight:
447.48
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥94.0% (acidimetric)
≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

-10 to -25°C

SMILES string

N(Cc4c(cc(cc4)OC)OC)(CC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C26H25NO6/c1-31-18-12-11-17(24(13-18)32-2)14-27(15-25(28)29)26(30)33-16-23-21-9-5-3-7-19(21)20-8-4-6-10-22(20)23/h3-13,23H,14-16H2,1-2H3,(H,28,29)

InChI key

UIDQSTVPYKMCEY-UHFFFAOYSA-N

General description

Fmoc-(Dmb)Gly-OH is an excellent reagent for enhancing synthetic efficiency of glycine-containing peptides by Fmoc SPPS .Like the analogous Hmb derivative Fmoc-(FmocHmb)Gly-OH, the use of this derivative prevents aggregation during chain assemby, thereby leading to faster and more predictable acylation and deprotection reactions. Furthermore, it can prevent aspartimide formation when used to introduce a Gly immediately before an Asp residue and help promote cyclization of Gly-containing peptides [1]. The analogous (Tmob)Gly derivative has been used by Bayer and colleagues to prepare a 64-residue transmembrane peptide in remarkable purity [2].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS

Literature references

[1] M. El Haddadi, et al. (2000) J. Pept. Sci., 6, 560 (2000) J. Pept. Sci., 6, 560.
[2] K. Jauch, et al. in "Peptides 1996: Proc. 24th European Peptide Symposium", R. Ramage and R. Epton (Eds), Mayflower Scientific Ltd., 1996, pp 497.

Linkage

Replaces: 04-12-1268

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(157B)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): passes test
Assay (acidimetric): ≥ 94.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service