Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52083

Sigma-Aldrich

Fmoc-Dpr(ivDde)-OH

≥99.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Dpr(ivDde)-OH, N-α-Fmoc-N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)-3-methylbutyl-L-diaminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H36N2O6
CAS Number:
Molecular Weight:
532.63
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Product Name

Fmoc-Dpr(ivDde)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥95.0% (acidimetric)
≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

SMILES string

N([C@@H](CNC(=C4C(=O)CC(CC4=O)(C)C)CC(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C31H36N2O6/c1-18(2)13-24(28-26(34)14-31(3,4)15-27(28)35)32-16-25(29(36)37)33-30(38)39-17-23-21-11-7-5-9-19(21)20-10-6-8-12-22(20)23/h5-12,18,23,25,32H,13-17H2,1-4H3,(H,33,38)(H,36,37)/t25-/m0/s1

InChI key

HLIFXCXTXPXGNH-VWLOTQADSA-N

General description

This orthogonally-protected diaminopropionic acid derivative is based on the hindered Dde variant ivDde. The side-chain ivDde group is considerably more stable to piperidine than Dde and is less prone to migrate from protected to unprotected side-chains [1]. However, migration from side-chain to the unprotected α-amino group of Dpr is unavoidable [2]. This side-reaction can, however, be minimized by the appropriate choice of coupling method for the subsequent residue, see [3].When removing ivDde in the presence of allyl-based protecting groups, allyl alcohol should be included in the deprotection solution to prevent reduction of the allyl group [4].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] S. R. Chhabra, et al. (1998) Tetrahedron Lett., 39, 1603.
[2] R. Wilhelm, et al., Poster 34 presented at the 16th American Peptide Symposium, Minneapolis, 1999.
[3] J. Beythien & P. Schneeberger, in ′Peptides 2000, Proc. 26th European Peptide Symposium′, EDK, Paris, 2001, pp. 361.
[4] B. Rohwedder, et al. (1998) Tetrahedron Lett., 39, 1175.

Linkage

Replaces: 04-12-1195

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(157A)): ≥ 98 %
Purity (TLC(CMA2)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 95.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service