Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52069

Sigma-Aldrich

Fmoc-Ser(PO(OBzl)OH)-OH

≥97% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Ser(PO(OBzl)OH)-OH, N-α-Fmoc-O-benzyl-L-phosphoserine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24NO8P
CAS Number:
Molecular Weight:
497.43
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Ser(PO(OBzl)OH)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥95.0% (HPLC)
≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C (−15°C to −25°C)

InChI

1S/C25H24NO8P/c27-24(28)23(16-34-35(30,31)33-14-17-8-2-1-3-9-17)26-25(29)32-15-22-20-12-6-4-10-18(20)19-11-5-7-13-21(19)22/h1-13,22-23H,14-16H2,(H,26,29)(H,27,28)(H,30,31)/t23-/m0/s1

InChI key

ZBPUWGDUVAAWJY-QHCPKHFHSA-N

General description

An excellent building block for the preparation of phosphoserine-containing peptides [1] by Fmoc SPPS. This derivative can be introduced using standard activation methods, such as PyBOP® and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods [2]. Applications of this derivative include the preparation of phospholamban [3], a 52 residue peptide containing both phosphoserine and phosphothreonine, and human salivary statherin, a 42 residue phosphoserine peptide [4]; for other examples see references [5,6,7,8].Recently, β-piperidinylalanine formation has been shown to occur during Fmoc deprotection of N-terminal Ser(PO(OBzl)OH), particularly under microwave conditions. This side reaction can be eliminated by using cyclohexylamine or DBU just for this Fmoc deprotection step [9].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] T. Wakamiya, et al. (1994) Chem. Lett., 1099.
[2] P. White & J. Beythien in ′Innovations & Perspectives in Solid Phase Synthesis and Combinatorial Libraries, 4th International Symposium′, Mayflower Scientific Ltd., Birmingham, 1996, pp. 557.
[3] H. Schmid, et al., Poster 423 presented at the 15th American Peptide Symposium, Nashville, 1997.
[4] T. L. Gururaja & M. J. Levine (1996) Pept. Res., 9, 283.
[5] T. Vorherr, et al. (1995) Bioorg. Med. Chem. Lett., 5, 2661.
[6] G. Shapiro, et al. (1996) Bioorg. Med. Chem. Lett., 6, 409.
[7] M. John, et al. (1996) Pept. Res., 9, 71.
[8] K. Teruya, et al. (2004) J. Pept. Sci., 10, 479.
[9] T. J. Attard, et al. (2009) Int. J. Pept. Res. Ther., 15, 69.

Linkage

Replaces: 04-12-1154

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(CMA1)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 1.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service