Skip to Content
MilliporeSigma

8.51064

mono-Fmoc ethylene diamine hydrochloride

≥98% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

mono-Fmoc ethylene diamine hydrochloride, Fmoc-NH(CH 2) 2NH 2 · HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18N2O2 · xHCl
CAS Number:
Molecular Weight:
282.34 (free base basis)
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

mono-Fmoc ethylene diamine hydrochloride, Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reagent type: spacer

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

Cl.N(CCN)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C17H18N2O2.ClH/c18-9-10-19-17(20)21-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16;/h1-8,16H,9-11,18H2,(H,19,20);1H

InChI key

UURYASDYOGIDRX-UHFFFAOYSA-N

General description

Ethylene diamine monoprotected with base-labile Fmoc protecting group.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA2)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0064

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library