Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51041

Sigma-Aldrich

Boc-NH-(PEG)₅-COOH (22 atoms)

Novabiochem®

Synonym(s):

Boc-NH-(PEG)₅-COOH (22 atoms), Boc-21-amino-4,7,10,13,16,19-hexaoxaheneicosanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H39NO10
CAS Number:
Molecular Weight:
453.52
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

oil

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N(CCOCCOCCOCCOCCOCCOCCC(=O)O)C(=O)OC(C)(C)C

InChI

1S/C20H39NO10/c1-20(2,3)31-19(24)21-5-7-26-9-11-28-13-15-30-17-16-29-14-12-27-10-8-25-6-4-18(22)23/h4-17H2,1-3H3,(H,21,24)(H,22,23)

InChI key

FTTYOIHYERRXQB-UHFFFAOYSA-N

General description

22-Atom long aminoacid PEG spacer. Can be introduced using Boc SPPS protocols

Application

Boc-NH-(PEG)₅-COOH (22 atoms) Novabiochem®. CAS 882847-13-4, molar mass 453.53 g/mol.

Linkage

Replaces: 01-63-0207

Analysis Note

Color (visual): colorless to slight yellow to yellow-brown
Appearance of substance (visual): oil
Identity (LC-MS): 452.2 - 454.2
Purity (TLC(011A)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service