Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43982

Sigma-Aldrich

Indium(III)-sulfate

anhydrous, for synthesis

Synonym(s):

Indium(III)-sulfate, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In2O12S3
CAS Number:
Molecular Weight:
517.82
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-689-1
NACRES:
NA.22
grade:
anhydrous
form:
solid

grade

anhydrous

Quality Level

form

solid

storage temp.

no temp limit

SMILES string

[In]2(O[S](=O)(=O)O2)O[S](=O)(=O)O[In]1O[S](=O)(=O)O1

InChI

1S/2In.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

XGCKLPDYTQRDTR-UHFFFAOYSA-H

General description

for synthesis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service