Skip to Content
MilliporeSigma

8.43969

Chlorohexidine

for synthesis

Synonym(s):

Chlorohexidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H30Cl2N10
CAS Number:
Molecular Weight:
505.45
UNSPSC Code:
12352101
EC Index Number:
200-238-7
NACRES:
NA.22
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

5000 mg/kg LD50, oral (Rat)

solubility

0.8 g/L

storage temp.

2-30°C

SMILES string

Clc1ccc(cc1)N\C(=N\C(=N)NCCCCCCNC(=N)\N=C(\Nc2ccc(cc2)Cl)/N)\N

InChI

1S/C22H30Cl2N10/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34)

InChI key

GHXZTYHSJHQHIJ-UHFFFAOYSA-N



pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library