Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43834

Sigma-Aldrich

1-Bromo-3-methoxypropane

for synthesis

Synonym(s):

1-Bromo-3-methoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9BrO
CAS Number:
Molecular Weight:
153.02
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

vapor pressure

6.6 hPa ( 20 °C)

Quality Level

Assay

≥97.5% (GC)

potency

930 mg/kg LD50, oral (Rat)

pH

6-7 ( in H2O)

bp

121 °C/1013 hPa

transition temp

flash point 43 °C

density

1.36 g/cm3

storage temp.

2-30°C

SMILES string

BrCCCOC

InChI key

CEVMYGZHEJSOHZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.355 - 1.365
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F

Flash Point(C)

43 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service