Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43818

Sigma-Aldrich

5-(4-Fluorophenyl)pyridine-3-carboxylic acid

for synthesis

Synonym(s):

5-(4-Fluorophenyl)pyridine-3-carboxylic acid, 5-(4-Fluorophenyl)nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8FNO2
CAS Number:
Molecular Weight:
217.20
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

pH

5.7 (20 °C, 5 g/L in H2O)

mp

215 °C

density

0.5 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Fc1ccc(cc1)c2cncc(c2)C(=O)O

InChI

1S/C12H8FNO2/c13-11-3-1-8(2-4-11)9-5-10(12(15)16)7-14-6-9/h1-7H,(H,15,16)

InChI key

KISDAKHYANOMTN-UHFFFAOYSA-N

Analysis Note

Colour (visual): white to offwhite
Appearance of substance (visual): powder
Assay (acidimetric): ≥ 97.0 %
Water (K. F.): ≤ 1.0 %
Melting range (lower value): ≥ 210 °C
Melting range (upper value): ≤ 215 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service