Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43785

Sigma-Aldrich

1-Phenylethyl acetate

for synthesis

Synonym(s):

1-Phenylethyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-288-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

450 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

212 °C/1013 hPa

mp

-60 °C

transition temp

flash point 91 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(C)c1ccccc1)C(=O)C

InChI

1S/C10H12O2/c1-8(12-9(2)11)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

QUMXDOLUJCHOAY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.024 - 1.027
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

198.5 °F

Flash Point(C)

92.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service