Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43483

Sigma-Aldrich

Linoleic acid

for synthesis

Synonym(s):

Linoleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-470-9
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

bp

230 °C/21 hPa

mp

-5 °C

transition temp

flash point >110 °C

density

0.903 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.900 - 0.910
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service