Skip to Content
MilliporeSigma

8.41535

4-Aminonaphthalene-1-sulfonic acid

for synthesis

Synonym(s):

4-Aminonaphthalene-1-sulfonic acid, 1-Naphthylamine-4-sulfonic acid, Naphthionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H9NO3S
CAS Number:
Molecular Weight:
223.25
UNSPSC Code:
12352106
EC Index Number:
201-567-9
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (acidimetric)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (acidimetric)

form

powder

mp

240-241 °C

density

1.67 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)c1c2c(c(cc1)N)cccc2

InChI

1S/C10H9NO3S/c11-9-5-6-10(15(12,13)14)8-4-2-1-3-7(8)9/h1-6H,11H2,(H,12,13,14)

InChI key

NRZRRZAVMCAKEP-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Identity (IR): passes test


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library