Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41467

Sigma-Aldrich

2-Bromoterephthalic acid

for synthesis

Synonym(s):

2-Bromoterephthalic acid, 2-Bromo-1,4-benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5BrO4
CAS Number:
Molecular Weight:
245.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
209-572-8
NACRES:
NA.22

Quality Level

Assay

≥97.0% (HPLC)

form

solid

mp

>300 °C

storage temp.

2-30°C

SMILES string

Brc1c(ccc(c1)C(=O)O)C(=O)O

InChI

1S/C8H5BrO4/c9-6-3-4(7(10)11)1-2-5(6)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

QPBGNSFASPVGTP-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service