Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41364

Sigma-Aldrich

n-Tricosane

for synthesis

Synonym(s):

n-Tricosane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48
CAS Number:
Molecular Weight:
324.63
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-347-4
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

flakes

autoignition temp.

524 °C

bp

199-200 °C/4 hPa

mp

48-49 °C

transition temp

flash point 113 °C

density

0.778 g/cm3 at 48 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCCCCCC)CCCCCCCCCCC

InChI

1S/C23H48/c1-3-5-7-9-11-13-15-17-19-21-23-22-20-18-16-14-12-10-8-6-4-2/h3-23H2,1-2H3

InChI key

FIGVVZUWCLSUEI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 47 °C
Melting range (upper value): ≤ 49 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service