Skip to Content
MilliporeSigma

8.41297

1,2-Benzenedimethanol

for synthesis

Synonym(s):

1,2-Benzenedimethanol, 1,2-Bishydroxymethylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
UNSPSC Code:
12352104
EC Index Number:
210-293-9
NACRES:
NA.22
MDL number:
Assay:
≥96.5% (GC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96.5% (GC)

form

powder

mp

62-64 °C

solubility

158 g/L

storage temp.

2-30°C

SMILES string

OCc1c(cccc1)CO

InChI

1S/C8H10O2/c9-5-7-3-1-2-4-8(7)6-10/h1-4,9-10H,5-6H2

InChI key

XMUZQOKACOLCSS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 63 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 1,2-Benzenedimethanol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library