Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.41225

Sigma-Aldrich

Methyl cyclopropanecarboxylate

for synthesis

Synonym(s):

Methyl cyclopropanecarboxylate, Cyclopropanecarboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352108
EC Index Number:
220-690-9
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

435 °C

bp

119 °C/1013 hPa

transition temp

flash point 17 °C

density

0.983 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H8O2/c1-7-5(6)4-2-3-4/h4H,2-3H2,1H3

InChI key

PKAHQJNJPDVTDP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.995 - 0.999
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Dermal - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service