Skip to Content

8.40104

1,5-Naphthalenedisulfonic acid

for synthesis

Synonym(s):

1,5-Naphthalenedisulfonic acid, Armstrong acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8O6S2
CAS Number:
Molecular Weight:
288.30
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-317-9
NACRES:
NA.22
Assay:
≥97.0% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

51 hPa ( 50 °C)

Quality Segment

assay

≥97.0% (HPLC)

potency

2420 mg/kg LD50, oral (Rat)

pH

2.4 (20 °C, 1 g/L in H2O)

mp

110 °C (decomposes)

solubility

1030 g/L

bulk density

900 kg/m3

SMILES string

[S](=O)(=O)([O-])c1c2c(c(ccc2)[S](=O)(=O)[O-])ccc1.[H+].[H+]

InChI

1S/C10H8O6S2/c11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16/h1-6H,(H,11,12,13)(H,14,15,16)

InChI key

XTEGVFVZDVNBPF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Water (K. F.): 22.0 - 26.0 %
Identity (IR): passes test


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

wgk

WGK 3

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library