Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.40037

Sigma-Aldrich

2,4-Dichlorobenzonitrile

for synthesis

Synonym(s):

2,4-Dichlorobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3Cl2N
CAS Number:
Molecular Weight:
172.01
MDL number:
UNSPSC Code:
12352117
EC Index Number:
229-493-2
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

crystals

mp

59-62 °C

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)C#N

InChI

1S/C7H3Cl2N/c8-6-2-1-5(4-10)7(9)3-6/h1-3H

InChI key

GRUHREVRSOOQJG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 59 °C
Melting range (upper value): ≤ 62 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service