Skip to Content
MilliporeSigma

8.22344

Acetanilide

for synthesis

Synonym(s):

Acetanilide, N-Phenylacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H9NO
CAS Number:
Molecular Weight:
135.16
UNSPSC Code:
12352111
EC Index Number:
203-150-7
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.002 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

solid

autoignition temp.

540 °C

potency

800 mg/kg LD50, oral (Rat)

pH

5-7 (25 °C, 10 g/L in H2O)

bp

304 °C/1013 hPa

mp

115 °C

transition temp

flash point 173 °C (DIN 51758)

solubility

5 g/L

density

1.22 g/cm3 at 20 °C

bulk density

370 kg/m3

storage temp.

2-30°C

SMILES string

N(c1ccccc1)C(=O)C

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Application

  • Adsorption Processes: Study on the removal of emerging pollutants, including acetanilide, through adsorption on activated carbons prepared by chemical activation of Organosolv lignin, relevant for environmental cleanup and material science (Gutiérrez-Martín et al., 2024).
  • Material Chemistry Applications: Utilization of acetanilide in the development of novel copper complexes, characterized for their potential application as heterogeneous catalysts for biodiesel production, linking organic synthesis with material science applications (Morad et al., 2020).

Analysis Note

Colour (visual): white to beige to brownish
Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 112 °C
Melting range (upper value): ≤ 115 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Acetanilide


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

321.8 °F - closed cup

flash_point_c

161 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library