Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22338

Sigma-Aldrich

Sulfanilic acid

for synthesis

Synonym(s):

Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3S
CAS Number:
Molecular Weight:
173.19
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-482-5
NACRES:
NA.22

grade

for synthesis
synthesis grade

Quality Level

Assay

≥99% substance (anhydrous) basis (acidimetric)

form

solid

autoignition temp.

>400 °C

potency

12300 mg/kg LD50, oral (Rat)

pH

2.5 (20 °C, 10 g/L in H2O)

mp

288 °C (decomposes)

solubility

water: 10 g/L at 20 °C

density

1.485 g/cm3 at 25 °C

bulk density

620 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)N

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric, calc. on anhydrous substance): ≥ 99.0 %
Loss on drying (110 °C 2h): ≤ 2.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service