Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22307

Sigma-Aldrich

Tetrahydrofurfuryl alcohol

for synthesis

Synonym(s):

Tetrahydrofurfuryl alcohol, Tetrahydrofurancarbinol, 2-Hydroxymethyltetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-625-6
NACRES:
NA.22

vapor pressure

1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

282 °C

potency

1600 mg/kg LD50, oral (Rat)

expl. lim.

1.5-9.7 % (v/v)

bp

178 °C/1013 hPa

mp

<-80 °C

transition temp

flash point 75 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(CCC1)CO

InChI

1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2

InChI key

BSYVTEYKTMYBMK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.052 - 1.054
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service