Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22267

Sigma-Aldrich

2,4,6-Trimethylpyridine

for synthesis

Synonym(s):

2,4,6-Trimethylpyridine, sym.-Collidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C5H2N
CAS Number:
Molecular Weight:
121.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-613-3
NACRES:
NA.22

vapor pressure

4 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

400 mg/kg LD50, oral (Rat)

bp

170-172 °C/1013 hPa

mp

-43 °C

transition temp

flash point 55 °C

solubility

35 g/L

density

0.914 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1c(cc(cc1C)C)C

InChI

1S/C8H11N/c1-6-4-7(2)9-8(3)5-6/h4-5H,1-3H3

InChI key

BWZVCCNYKMEVEX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.913 - 0.915
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service