Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

8.22265

Supelco

Chloroform

EMPLURA®

Synonym(s):

TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-663-8
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

potency

695 mg/kg LD50, oral (Rat)
>3980 mg/kg LD50, skin (Rabbit)

impurities

≤0.1% Water

evapn. residue

≤0.0010%

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Improved lipid analysis using a 2D-LC-MS system with a novel injection procedure.: This research introduces an advanced lipid analysis method that incorporates chloroform in a dual-dimensional liquid chromatography-mass spectrometry setup, enhancing lipidomic studies′ precision and throughput (Schiell et al., 2024).

  • Uncovering the Power of HSCCC: Separation of Diastereomeric and Regioisomeric Styrylpyrones from the Stem Bark of Goniothalamus leiocarpus.: Describes the use of chloroform in high-speed countercurrent chromatography, which is critical for separating complex natural product mixtures (Mu et al., 2024).

  • Determination of aflatoxins in rice from Penang, Malaysia by dispersive liquid-liquid micro-extraction and LC-MS/MS.: This study employs chloroform in a dispersive liquid-liquid micro-extraction technique for detecting aflatoxins in rice, highlighting its role in food safety (Ismail et al., 2024).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 1.474 - 1.479
Evaporation residue: ≤ 0.0010 %
Water: ≤ 0.10 %
Stabilized with 0,7 - 1,0% Ethanol

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service