Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22261

Sigma-Aldrich

Pyrocatechol

for synthesis

Synonym(s):

Pyrocatechol, 1,2-Dihydroxybenzene, Catechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-427-5
NACRES:
NA.22

vapor pressure

13 hPa ( 118 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

potency

300 mg/kg LD50, oral (Rat)

expl. lim.

1.97 % (v/v)

pH

6 (20 °C, 100 g/L in H2O)

mp

103-105 °C

transition temp

flash point 127 °C

solubility

430 g/L

density

1.37 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(cccc1)O

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H

InChI key

YCIMNLLNPGFGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pyrocatechol alleviates cisplatin-induced acute kidney injury by inhibiting ROS production: Research on pyrocatechol′s protective effects against nephrotoxicity due to cisplatin, highlighting its antioxidant properties (Xie et al., 2022).

  • How was the proton transfer process in bis-3, 6-(2-benzoxazolyl)-pyrocatechol, single or double proton transfer: Theoretical insights into the proton transfer mechanisms in a symmetric pyrocatechol derivative, providing fundamental knowledge for molecular interactions (Zhang et al., 2016).

  • Electrodeposition of pyrocatechol based films: Influence of potential scan rate, pyrocatechol concentration and pH: Investigates the conditions affecting the electrodeposition of pyrocatechol-based materials, contributing to the understanding of electrochemical coating processes (Ball, 2017).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 103 °C
Melting range (upper value): ≤ 105 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service