Skip to Content

8.22147

Tetraethylammonium bromide

greener alternative

Greener Alternative

for synthesis

Synonym(s):

Tetraethylammonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C2H5)4NBr
CAS Number:
Molecular Weight:
210.16
UNSPSC Code:
12352107
EC Index Number:
200-769-4
NACRES:
NA.22
MDL number:
Assay:
≥ 99.0%, ≥99% (argentometric)
Grade:
synthesis grade
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

assay

≥ 99.0%, ≥99% (argentometric)

form

crystals

reaction suitability

core: ammonium

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

dilution

(for synthesis)

pH

6.5 (20 °C, 100 g/L in H2O)

mp

>285 °C

density

1.397 g/cm3 at 20 °C

greener alternative category

storage temp.

2-30°C

SMILES string

[Br-].[N+](CC)(CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This advanced-grade supporting electrolyte is used for electrochemical applications, enabling efficient charge transfer and enhanced performance, offering a greener, potentially more sustainable option for specialized electrochemical processes. Click here for more information.

Application

  • Single-Atom Catalysts: Utilized in the synthesis of surfactant encapsulated palladium-polyoxometalates, TEAB was instrumental in controlled assembly processes, demonstrating potential in catalytic applications and material sciences for drug delivery systems (He et al., 2016).
  • All Silica Zeolite Synthesis: TEAB was used as a template in the solvent-free synthesis of all-silica beta zeolite, showing significance in the industrial application of silica materials, which are critical in various filtering and catalytic operations in material sciences (Cui et al., 2018).
  • Wide Color Gamut Backlit Display: Research highlighted TEAB′s role in the development of high quantum efficiency zinc-doped lead manganese halide films for backlight display applications, illustrating its potential in advanced functional materials used in display technologies (Yang et al., 2023).
  • Interfacial Engineering of Emitter Films: TEAB was incorporated to modulate grain size and enhance the interfacial properties of CH3NH3PbBr3 emitter films, suggesting its application in improving photovoltaic and display technologies, relevant for material science and renewable energy sectors (Jamaludin et al., 2018).
  • X-Ray Scintillation Applications: Applied in the synthesis of environmentally friendly manganese halides for X-ray scintillation, TEAB supported advancements in medical imaging technologies, potentially useful in drug discovery and diagnostic applications (Jiang et al., 2021).

Analysis Note

Assay (argentometric): ≥ 99.0 %
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Tetraethylammonium bromide


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

361.4 °F - closed cup

flash_point_c

183 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library