Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22110

Sigma-Aldrich

n-Nonadecane

for synthesis

Synonym(s):

n-Nonadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17CH3
CAS Number:
Molecular Weight:
268.52
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-116-8
NACRES:
NA.22
grade:
for synthesis
bp:
329-331 °C
vapor pressure:
<0.1 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

solid

autoignition temp.

205 °C

expl. lim.

0.6-6.5 % (v/v)

dilution

(for synthesis)

bp

329-331 °C

mp

31-34 °C

transition temp

flash point >100 °C

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCCCC)CCCCCCCCC

InChI

1S/C19H40/c1-3-5-7-9-11-13-15-17-19-18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3

InChI key

LQERIDTXQFOHKA-UHFFFAOYSA-N

Application


  • Epoxy⁻PCM Composites with Nanocarbons or Multidimensional Boron Nitride as Heat Flow Enhancers.: This study explores the application of epoxy⁻phase change material (PCM) composites integrated with nanocarbons or multidimensional boron nitride to enhance heat flow properties. n-Nonadecane is used as a PCM due to its suitable thermal properties, demonstrating its potential in improving the efficiency of thermal management systems (Agrawal et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 31 °C
Melting range (upper value): ≤ 34 °C
Identity (GC): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service