Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.22047

Sigma-Aldrich

1-Docosanol

for synthesis

Synonym(s):

1-Docosanol, Behenyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46O
CAS Number:
Molecular Weight:
326.60
MDL number:
UNSPSC Code:
12352104
EC Index Number:
211-546-6
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

solid

bp

180 °C/0.3 hPa

mp

68-71 °C

storage temp.

2-30°C

InChI

1S/C22H46O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3

InChI key

NOPFSRXAKWQILS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 71 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

410.0 °F

Flash Point(C)

210 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service