Skip to Content
MilliporeSigma

8.21940

Ethylenediamine-N,N,N′,N′-tetra-2-propanol

for synthesis

Synonym(s):

Ethylenediamine-N,N,N′,N′-tetra-2-propanol, EDTP, N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3CH(OH)CH2]2NCH2CH2N[CH2CH(OH)CH3]2
CAS Number:
Molecular Weight:
292.41
UNSPSC Code:
12352104
EC Index Number:
203-041-4
NACRES:
NA.22
MDL number:
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

pH

10.4 (20 °C, 10 g/L in H2O)

bp

240 °C/13 hPa

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCN(CC(O)C)CC(O)C)(CC(O)C)CC(O)C

InChI

1S/C14H32N2O4/c1-11(17)7-15(8-12(2)18)5-6-16(9-13(3)19)10-14(4)20/h11-14,17-20H,5-10H2,1-4H3

InChI key

NSOXQYCFHDMMGV-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

411.8 °F - closed cup

flash_point_c

211 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library