Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21083

Sigma-Aldrich

Tetraethyl orthotitanate

for synthesis

Synonym(s):

Tetraethyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)4Ti
CAS Number:
Molecular Weight:
228.11
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-410-8
NACRES:
NA.22

Quality Level

form

liquid

bp

150-155 °C/16 hPa

mp

-40 °C

transition temp

flash point 33 °C

density

1.08 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ti+4].[O-]CC.[O-]CC.[O-]CC.[O-]CC

InChI

1S/4C2H5O.Ti/c4*1-2-3;/h4*2H2,1H3;/q4*-1;+4

InChI key

JMXKSZRRTHPKDL-UHFFFAOYSA-N

Analysis Note

Assay (ex Ti): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.084 - 1.096
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service