Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

8.20820

Supelco

Isobutyl methyl ketone

EMPLURA®

Synonym(s):

4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-550-1
NACRES:
NA.05
bp:
117-118 °C
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

840 °F

potency

2080 mg/kg LD50, oral (Rat)
>16000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-8 %, 93 °F

dilution

(low-cost alternative, for basic applications)

refractive index

n20/D 1.395 (lit.)

pH

7 (20 °C)

bp

117-118 °C

mp

−80 °C (lit.)

transition temp

flash point 14 °C

solubility

14.1 g/L

density

0.801 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.800 - 0.802

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service