Skip to Content
MilliporeSigma

8.20696

2-Imidazolidinethione

for synthesis

Synonym(s):

2-Imidazolidinethione, N,N′-Ethylenethiourea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
UNSPSC Code:
12352115
EC Index Number:
202-506-9
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<1 hPa ( 25 °C)

Quality Segment

assay

≥98.0% (HPLC)

form

solid

potency

710 mg/kg LD50, oral (Rat)

bp

240 °C/1010 hPa

mp

199 °C

transition temp

flash point 253 °C

solubility

8 g/L

density

1.4 g/cm3 at 20 °C

bulk density

750‑900 kg/m3

storage temp.

2-30°C

SMILES string

S=C1NCCN1

InChI

1S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

PDQAZBWRQCGBEV-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 196 °C
Melting range (upper value): ≤ 199 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 2-Imidazolidinethione


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library