Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20624

Sigma-Aldrich

1-Heptanol

for synthesis

Synonym(s):

1-Heptanol, Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-897-9
NACRES:
NA.22

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

350 °C

potency

500 mg/kg LD50, oral (Rat)
2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9 % (v/v)

bp

176 °C/1013 hPa

mp

-35 °C

transition temp

flash point 78 °C

solubility

1 g/L

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.822 - 0.823
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service