Skip to Content
MilliporeSigma

8.20604

Glutaric anhydride

for synthesis

Synonym(s):

Glutaric anhydride, Tetrahydropyran-2,6-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
UNSPSC Code:
12352115
EC Index Number:
203-593-6
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.05 hPa ( 50 °C)

Quality Level

form

solid

autoignition temp.

320 °C

potency

1780 mg/kg LD50, skin (Rabbit)

bp

290.5 °C/1013 hPa

mp

53-57 °C

transition temp

flash point 175 °C

density

1.24 g/cm3 at 60 °C

bulk density

990 kg/m3

storage temp.

2-30°C

SMILES string

O1C(=O)CCCC1=O

InChI

1S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2

InChI key

VANNPISTIUFMLH-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 95.0 %
Melting range (lower value): ≥ 53 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Glutaric anhydride


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

347.0 °F

flash_point_c

175 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
820604025004022536462554