Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20565

Sigma-Aldrich

2-Fluoroaniline

for synthesis

Synonym(s):

2-Fluoroaniline, 1-Amino-2-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(F)C6H4NH2
CAS Number:
Molecular Weight:
111.12
MDL number:
UNSPSC Code:
12352116
EC Index Number:
206-478-9
NACRES:
NA.22

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

171-173 °C/1013 hPa

mp

-29 °C

transition temp

flash point 60 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H6FN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

FTZQXOJYPFINKJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.150 - 1.153
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service