Skip to Content
MilliporeSigma

8.20543

n-Dodecane

for synthesis

Synonym(s):

n-Dodecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
UNSPSC Code:
12352001
EC Index Number:
203-967-9
NACRES:
NA.22
MDL number:
Assay:
≥99% (GC)
Grade:
synthesis grade
Bp:
216.3 °C/1013 hPa
Vapor pressure:
0.4 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.4 hPa ( 20 °C)

assay

≥99% (GC)

form

liquid

autoignition temp.

200 °C

expl. lim.

0.6 % (v/v)

dilution

(for synthesis)

kinematic viscosity

<7 cSt(40 °C)

bp

216.3 °C/1013 hPa

mp

-9.6 °C

transition temp

flash point 85.5 °C

density

0.75 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCC)CCCCC

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

General description

n-Dodecane is used as a solvent in organic chemistry. It is used as a distillation chaser and scintillator component. Currently, n-dodecane is being considered an alternative to kerosene-based fuels.

Application

n-Dodecane is used as a:
  • Non-polar solvent for the direct synthesis of indium nanoparticles.
  • Diluent in bifunctional ionic liquid (IL) to extract lithium from brines.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.748 - 0.749
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under n-Dodecane


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

flash_point_f

158.0 °F - Pensky-Martens closed cup

flash_point_c

70 °C - Pensky-Martens closed cup

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library