Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

820464

Sigma-Aldrich

Probumin® Bovine Serum Albumin Diagnostic Grade

29-31 g/dL protein (Biuret), USA origin, Tested for protease, enzyme immunoassay: suitable, ELISA: suitable, Serology: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801

Product Name

Probumin® Bovine Serum Albumin Diagnostic Grade, solution (30%), pkg of 50 mL

Assay

29-31 g/dL protein basis (biuret)

Quality Level

form

solution (30%)

shelf life

48 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pkg of 50 mL

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL protein (biuret)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

dilution

(Serology)

impurities

Tested for IgG

pH

7.2-7.4

foreign activity

Bluetongue, none detected
Tested for Protease
Tested for protease
Vesicular Stomatitis Virus, none detected
microbial content by USP tested
preservative, free

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Related Content

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service