Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.20451

Sigma-Aldrich

2,4-Dichlorophenoxyacetic acid

for synthesis

Synonym(s):

2,4-Dichlorophenoxyacetic acid, 2,4 D-acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,4-(Cl)2C6H3OCH2COOH
CAS Number:
Molecular Weight:
221.04
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-361-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa

Quality Level

200
300

Assay

≥98.0% (acidimetric)

form

solid

potency

375 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

160 °C/0.53 hPa

mp

134-137 °C

solubility

0.3 g/L

density

1.42 g/cm3 at 25 °C

bulk density

720 kg/m3

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)OCC(=O)O

InChI

1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

OVSKIKFHRZPJSS-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 134 °C
Melting range (upper value): ≤ 140 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service