Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20248

Sigma-Aldrich

Di-tert-butyl peroxide

for synthesis

Synonym(s):

Di-tert-butyl peroxide, tert-Butyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18O2
CAS Number:
Molecular Weight:
146.23
MDL number:
UNSPSC Code:
12352120
EC Index Number:
203-733-6
NACRES:
NA.22

vapor pressure

1000 hPa ( 110 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

182 °C

potency

>25000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

mp

-40 °C

transition temp

flash point 4 °C

density

0.80 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C8H18O2/c1-7(2,3)9-10-8(4,5)6/h1-6H3

InChI key

LSXWFXONGKSEMY-UHFFFAOYSA-N

Application


  • Combustion characteristics of DTBP: Investigates the combustion characteristics of DTBP, emphasizing its role as a cetane booster and a source for methyl radicals, which are essential in studying elementary reactions (Preubker et al., 2021).

  • DTBP in alpha-Alkylation of alpha-Amino Carbonyl Compounds: Details a DTBP-promoted alpha-alkylation process that enhances the synthesis efficiency of alpha-amino carbonyl compounds by simple alkanes (Peng et al., 2014).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.795 - 0.797
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Muta. 2 - Org. Perox. E

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service