Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20170

Sigma-Aldrich

Pyruvic acid

for synthesis

Synonym(s):

Pyruvic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-824-3
NACRES:
NA.22

Quality Level

Assay

≥98.0% (acidimetric)

form

liquid

autoignition temp.

305 °C

pH

1.2 (20 °C, 90 g/L in H2O)

mp

12 °C

transition temp

flash point 82-83 °C

density

1.27 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

OC(=O)C(=O)C

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)

InChI key

LCTONWCANYUPML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Semiconducting polymer dots based L-lactate sensor: This study outlines the development of a semiconducting polymer dots-based L-lactate sensor utilizing an enzymatic cascade reaction system including pyruvic acid, illustrating its application in biochemical assays (He et al., 2024).
  • Antioxidant activity and metabolomic analysis: Research on Streptococcus alactolyticus included pyruvic acid in its metabolomic profiling to assess antioxidant activities, showcasing its importance in microbial metabolism studies (Gu et al., 2024).
  • Sodium alginate hydrogelation mediated paper-based POCT sensor: This paper introduces a sodium alginate hydrogelation-mediated sensor for dual-signal determination of L-lactate, incorporating pyruvic acid as a key reactant, suitable for point-of-care testing applications (Wang et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.269 - 1.276
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service