Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20128

Sigma-Aldrich

Saccharin

for synthesis

Synonym(s):

Saccharin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-321-0
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (acidimetric)

form

crystals

mp

228-230 °C

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)NC(=O)c2c1cccc2

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Application


  • Gabapentin-saccharin co-crystals with enhanced physicochemical properties and in vivo absorption formulated as oro-dispersible tablets.: This study presents the formulation of gabapentin-saccharin co-crystals, demonstrating improved solubility and absorption, beneficial for pharmaceutical applications (Soliman et al., 2020, doi: 10.1080/10837450.2019.1687521).

  • Improving the solubility, dissolution, and bioavailability of Ibrutinib by preparing it in a coamorphous state with saccharin.: The research focuses on enhancing the bioavailability of Ibrutinib through coamorphous preparation with saccharin, which is significant for cancer treatment (Shi et al., 2019, doi: 10.1016/j.xphs.2019.04.031).


Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 226 °C
Melting range (upper value): ≤ 230 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service