Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.18685

Sigma-Aldrich

(2S,3S)-(-)-Diethyl tartrate

for synthesis

Synonym(s):

(2S,3S)-(-)-Diethyl tartrate, L-(+)-Tartaric acid diethyl ester, DET, (-)-Diethyl-D-tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OOC(CHOH)2COOC2H5
CAS Number:
Molecular Weight:
206.19
MDL number:
UNSPSC Code:
12352108
EC Index Number:
237-458-8
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

162 °C/25 hPa

mp

17 °C

transition temp

flash point 93 °C

density

1.20 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O[C@@H]([C@H](O)C(=O)OCC)C(=O)OCC

InChI

1S/C8H14O6/c1-3-13-7(11)5(9)6(10)8(12)14-4-2/h5-6,9-10H,3-4H2,1-2H3/t5-,6-/m0/s1

InChI key

YSAVZVORKRDODB-WDSKDSINSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.203 - 1.205
Spec. rotation [α²0/D (c=1 in water): -27.0 - -24.0 °
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service