Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.18550

Sigma-Aldrich

3-(Trimethoxysilyl)-propylamine

for synthesis

Synonym(s):

3-(Trimethoxysilyl)-propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17NO3Si
CAS Number:
Molecular Weight:
179.29
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-511-5
NACRES:
NA.22

Quality Level

form

liquid

autoignition temp.

295 °C

pH

>9 (20 °C, 20 g/L in H2O)

bp

194 °C/1013 hPa

transition temp

flash point 90 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H17NO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-7H2,1-3H3

InChI key

SJECZPVISLOESU-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.013 - 1.016
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service