Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18537

Sigma-Aldrich

4-Fluorobenzaldehyde

for synthesis

Synonym(s):

4-Fluorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(F)C6H4CHO
CAS Number:
Molecular Weight:
124.11
MDL number:
UNSPSC Code:
12352114
EC Index Number:
207-293-6

vapor pressure

19 hPa ( 70 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

70-72 °C/19 hPa

mp

-10 °C

transition temp

flash point 62 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H5FO/c8-7-3-1-6(5-9)2-4-7/h1-5H

InChI key

UOQXIWFBQSVDPP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.176 - 1.180
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Zhao et al.
Journal of agricultural and food chemistry, 68(1), 185-192 (2019-12-10)
Phytopathogenic fungi have been considered as being an enormous threat in the agricultural system. In our search of new antifungal natural products, nine new halogenated cyclopentenones, bicolorins A-I (1-3, and 5-10), along with three known cyclopentenones (4, 11, and 12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service