Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18383

Sigma-Aldrich

Amantadine sulfate

for synthesis

Synonym(s):

Amantadine sulfate, 1-Aminoadamantane sulfate, Adamantylamine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34N2 · xH2SO4
CAS Number:
Molecular Weight:
302.50 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
250-604-5
NACRES:
NA.22

Quality Level

pH

4.9 (20 °C, 100 g/L in H2O, slurry)

mp

299-301 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+H3]C54CC6CC(C5)CC(C4)C6.[N+H3]C21CC3CC(C2)CC(C1)C3

InChI

1S/2C10H17N.H2O4S/c2*11-10-4-7-1-8(5-10)3-9(2-7)6-10;1-5(2,3)4/h2*7-9H,1-6,11H2;(H2,1,2,3,4)

InChI key

MYWTWSQFJLXGGQ-UHFFFAOYSA-N

Analysis Note

Assay (of S): ≥ 98.0 %
Sulfated ash:
≤ 0.20 %
Identity (IR):
passes test
Sulfated ash:
≤ 0.20 %
Chloride: ≤ 0.04 %
Identity (IR):
passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service