Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18247

Sigma-Aldrich

Copper(II) chloride

for synthesis

Synonym(s):

Copper(II) chloride, Copper dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Cu
CAS Number:
Molecular Weight:
134.45
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-210-2
NACRES:
NA.22

Quality Level

form

solid

potency

584 mg/kg LD50, oral (Rat)

pH

3.5 (20 °C, 50 g/L in H2O)

mp

498 °C (decomposes)

solubility

620 g/L

density

3.39 g/cm3 at 20 °C

bulk density

1200 kg/m3

storage temp.

2-30°C

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Application

  • Catalytic enhancement in organic synthesis: The study explores the use of Copper(II) chloride as a catalytic agent in the synthesis of 4-sulfonyl-1,2,3-triazoles, highlighting its role in sequential aerobic copper-catalyzed sulfonylation and Dimroth cyclization, providing a robust method for constructing triazole frameworks in medicinal chemistry and material science (Van Hoof et al., 2021).

Analysis Note

Assay (iodometric, calc.on anhydrous substance): ≥ 98.0 %
Loss on drying (250 °C, 2 h): ≤ 5.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service