Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18213

Sigma-Aldrich

tert-Butyl glycinate

for synthesis

Synonym(s):

tert-Butyl glycinate, Glycine tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2COOC(CH3)3
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22

Quality Level

Assay

≥92.5% (GC)

form

liquid

bp

29-31 °C/3 hPa

density

0.96 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

NCC(=O)OC(C)(C)C

InChI

1S/C6H13NO2/c1-6(2,3)9-5(8)4-7/h4,7H2,1-3H3

InChI key

SJMDMGHPMLKLHQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 92.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.959 - 0.962
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service