Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18164

Sigma-Aldrich

Tetra-n-butylammonium fluoride trihydrate

for synthesis

Synonym(s):

Tetra-n-butylammonium fluoride trihydrate, TBAF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4NF · 3H2O
CAS Number:
Molecular Weight:
315.51
MDL number:
UNSPSC Code:
12352107
EC Index Number:
207-057-2
NACRES:
NA.22

Quality Level

form

crystals

mp

62-63 °C

storage temp.

2-30°C

SMILES string

[F-].[N+](CCCC)(CCCC)(CCCC)CCCC.O.O.O

InChI

1S/C16H36N.FH.3H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;/h5-16H2,1-4H3;1H;3*1H2/q+1;;;;/p-1

InChI key

VEPTXBCIDSFGBF-UHFFFAOYSA-M

Application

  • Unveiling the Use of 1,1-Bis(triflyl)ethylene as CF(3)SO(2)CH-CH(2) Source with the Assistance of (n-Bu)(4)NF.: This study discusses a synthetic approach using Tetra-n-butylammonium fluoride (TBAF) for generating 3-[(Trifluoromethyl)sulfonyl]cyclobut-1-enes, highlighting its role in mediating fluoride ion transfer in organic synthesis (Almendros et al., 2024).
  • Speciation and kinetics of fluoride transfer from tetra-n-butylammonium difluorotriphenylsilicate (′TBAT′).: The article explores the speciation and kinetics involved in fluoride transfer processes using TBAT, a compound closely related to TBAF, providing insights into its utility in synthetic chemistry (Lloyd-Jones et al., 2024).
  • Conjugated Polymers with Self-Immolative Sidechain Linkers for Carbon Nanotube Dispersion.: This research utilizes TBAF in the development of novel conjugated polymers with self-immolative sidechains for effective dispersion of carbon nanotubes, demonstrating its application in materials science and nanotechnology (Adronov et al., 2024).
  • Room temperature synthesis of CdSe/CdS triangular nanoemitters and their stabilization in colloidal state and sol-gel glass.: The article discusses the synthesis of semiconductor nanostructures at room temperature, where TBAF plays a critical role in the stabilization of colloidal solutions (Parola et al., 2023).

Analysis Note

Assay (titration): ≥ 97.0 %


Water (K. F.): 15.0 - 19.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service